Learn More
TMED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14141-1-AP-150UL
 View more versions of this product
Product Code. 16882644
  • 4652.90 SEK / 150µL
Description
Involved in endoplasmic reticulum stress response. May play a role in the regulation of heat-shock response and apoptosis.
Specifications
Specifications
TMED4
Polyclonal
Unconjugated
TMED4
ERS25, HNLF, TMED4
Rabbit
Antigen Affinity Chromatography
RUO
103694, 222068, 305502
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z7H5, Q8R1V4
Tmed4
TMED4 Fusion Protein Ag5234
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TMED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated