missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15320-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.Specifications
TNFAIP1 | |
Polyclonal | |
Unconjugated | |
TNFAIP1 | |
B12, B61, BACURD2, EDP1, hBACURD2, Protein B12, TNFAIP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21927, 287543, 7126 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13829, Q7TNY1 | |
Tnfaip1 | |
TNFAIP1 Fusion Protein Ag0220 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TNFAIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction