missing translation for 'onlineSavingsMsg'
Learn More

TPPP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13633-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881944

  • 1355.00 SEK / 20µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TPPP2
Polyclonal
Unconjugated
TPPP2
C14orf8, P18, p25beta, Protein p25 beta, TPPP/p18, TPPP2
Rabbit
Antigen Affinity Chromatography
RUO
122664, 219038, 498497
-20°C
Liquid
Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59282, Q0P5Y3
Tppp2
TPPP2 Fusion Protein Ag4561
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers