Learn More
TPST1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12404-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890104
  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.
Description
Description
The TPST1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TPST1. This antibody recognizes human, mouse, rat antigen. The TPST1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TPST1
Polyclonal
Unconjugated
20 μL
8460
IgG
ELISA, Immunohistochemistry, Western Blot
233 μg/mL
Rabbit
Primary
Mouse, Rat, Human