missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPT1 Mouse anti-Human, Mouse, Rat, Clone: 1G6A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66713-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody detected 22 kDa TPT1 in lysates.
TPT1 (translationally controlled tumor protein 1) is a 23 kDa member of the TCTP family of calcium binding proteins. TPT1 is localized in the cytoplasm and widely expressed and serves as a transcriptional activator, calcium transporter, histamine inducer and antiapoptotic caspase 3 inhibitor. TPT1 is a cytokine-like molecule that causes the release of histamine, IL-4 and IL-13 from basophils as well as the secretion of IL-8 and a calcium response in eosinophils. TPT1 plays a pivotal role in allergic diseases and due to its wide distribution in brain, is thought to be involved in neurodegenerative disorders, such as Alzheimer′s disease and Down syndrome.Specifications
TPT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13693, P63028, P63029 | |
TPT1 | |
TPT1 Fusion Protein Ag27651 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
1G6A10 | |
Unconjugated | |
TPT1 | |
FLJ27337, Fortilin, Histamine releasing factor, HRF, p02, p23, TCTP, TPT1 | |
Mouse | |
Protein G | |
RUO | |
116646, 22070, 7178 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TPT1 Mouse anti-Human, Mouse, Rat, Clone: 1G6A10, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction