missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC9/NIBP Mouse anti-Human, Mouse, Clone: 1C4F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66131-1-IG-20UL
This item is not returnable.
View return policy
Description
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.Specifications
TRAPPC9/NIBP | |
Monoclonal | |
1.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3U0M1, Q96Q05 | |
Trappc9 | |
TRAPPC9,NIBP Fusion Protein Ag8949 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin) | |
1C4F9 | |
Unconjugated | |
TRAPPC9 | |
IBP, IKBKBBP, KIAA1882, NIBP, NIK and IKBKB binding protein, T1, TRAPPC9, TRS120, Tularik gene 1 protein | |
Mouse | |
Protein A | |
RUO | |
76510, 83696 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRAPPC9/NIBP Mouse anti-Human, Mouse, Clone: 1C4F9, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction