missing translation for 'onlineSavingsMsg'
Learn More

TRAPPC9/NIBP Mouse anti-Human, Mouse, Clone: 1C4F9, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen TRAPPC9/NIBP
Clone 1C4F9
Concentration 1.8 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897725
View Documents
Proteintech
66131-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16887725
View Documents
Proteintech
66131-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.
Specifications

Specifications

TRAPPC9/NIBP
1.8 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
IBP, IKBKBBP, KIAA1882, NIBP, NIK and IKBKB binding protein, T1, TRAPPC9, TRS120, Tularik gene 1 protein
Trappc9
IgG2b
Protein A
Antibody
1C4F9
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Mouse, Human
Q3U0M1, Q96Q05
76510, 83696
TRAPPC9,NIBP Fusion Protein Ag8949
Primary
-20°C
TRAPPC9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TRAPPC9/NIBP Mouse anti-Human, Mouse, Clone: 1C4F9, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.