Learn More
Cytiva Tray Support for Gel Stainer and Processor Plus
Brand: Cytiva 80-6344-29
This item is not returnable.
View return policy
Description
Specifications
Gel Stainer and Processor Plus, Blot Processor | |
Tray Support | |
4.22 oz. | |
125 mL |
Product Content Correction
Product Title
Cytiva Tray Support for Gel Stainer and Processor Plus > 125mL
Spot an opportunity for improvement?Share a Content Correction