Learn More
Triiron dodecacarbonyl, 99%, stabilized Product Code.: 10115212
Triiron dodecacarbonyl, 99%, stabilized
Product Code. p-3213288 Shop All Thermo Scientific Chemicals Products
5 g, Glass bottle, 5g
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. 10115212
Brand: Thermo Scientific Acros 209290050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 17685-52-8 | C12Fe3O12 | 503.66 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17685-52-8
Molecular Formula C12Fe3O12
Molecular Weight (g/mol) 503.66
MDL Number MFCD00135617
Synonym triiron dodecarbonyl
Specifications
Melting Point 140.0°C
Color Black
Density 1.9900g/mL
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Fe3(CO)12
Quantity 5 g
Fieser 04,534; 05,706; 07,144; 08,41
Specific Gravity 1.99
Solubility Information Solubility in water: insoluble
Stabilizer 5 to 10% MeOH
Formula Weight 503.66
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Triiron dodecacarbonyl
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triiron dodecacarbonyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H371-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Product Title
Triiron dodecacarbonyl, 99%, stabilized >