Learn More
TRIM5 Mouse anti-Human, Unconjugated, Clone: 1H1G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67492-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The TRIM5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM5. This antibody recognizes human antigen. The TRIM5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TRIM5 | |
Monoclonal | |
660 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1H1G9 | |
Unconjugated | |
20 μL | |
85363 | |
IgG2b |