missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM5 Mouse anti-Human, Unconjugated, Clone: 1H1G9, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TRIM5 |
---|---|
Clone | 1H1G9 |
Concentration | 660 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800156
|
Proteintech
67492-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890146
|
Proteintech
67492-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TRIM5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM5. This antibody recognizes human antigen. The TRIM5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TRIM5 | |
660 ÎĽg/mL | |
Monoclonal | |
Mouse | |
85363 | |
Primary |
1H1G9 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRIM5 Mouse anti-Human, Unconjugated, Clone: 1H1G9, Proteintech