Learn More
TRIM5 Mouse anti-Human, Unconjugated, Clone: 1H1G9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67492-1-IG-20UL
 View more versions of this product
Product Code. 16800156
  • 1645.54 SEK / 20µL
Description
The TRIM5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM5. This antibody recognizes human antigen. The TRIM5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRIM5
Monoclonal
660 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1H1G9
Unconjugated
20 μL
85363
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TRIM5 Mouse anti-Human, Unconjugated, Clone: 1H1G9, Proteintech > 20 μL