Learn More
Proteintech TRIM5 Mouse anti-Human, Unconjugated, Clone: 1H1G9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67492-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890146
  • 3665.00 SEK / 150µL
Estimated Shipment: 08-06-2023
to see stock.
Description
Description
The TRIM5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM5. This antibody recognizes human antigen. The TRIM5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRIM5
Monoclonal
660 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1H1G9
Unconjugated
150 μL
85363
IgG2b