missing translation for 'onlineSavingsMsg'
Learn More
Trimethyltin hydroxide, 98% Product Code.: 15234764

Trimethyltin hydroxide, 98%

Product Code. 15234764 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle, 25g
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15234764

Brand: Thermo Scientific Acros 449220250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 56-24-6 | C3H10OSn | 180.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-24-6
Molecular Formula C3H10OSn
Molecular Weight (g/mol) 180.82
MDL Number MFCD00013929
InChI Key OJZNYUDKNVNEMV-UHFFFAOYSA-M
Synonym trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide
PubChem CID 6328620
IUPAC Name trimethyltin;hydrate
SMILES C[Sn](C)(C)O

Specifications

Melting Point 118.0°C to 119.0°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3SnOH
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 180.82
Percent Purity 98%
Chemical Name or Material Trimethyltin hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0451618 Certificate TypeCertificate of Analysis Date02/05/2023 Product Code449220050,449220250,449220010
Lot NumberA0409527 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code449220050,449220250,449220010
Lot NumberA0383496 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code449220050,449220250,449220010
Lot NumberA0416022 Certificate TypeCertificate of Analysis Date12/12/2022 Product Code449220050,449220250,449220010
Lot NumberA0436845 Certificate TypeCertificate of Analysis Date22/12/2021 Product Code449220050,449220250,449220010
compliance-icons compliance-icons
Product Identifier
  • Trimethyltin hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethyltin hydroxide, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.