missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT61B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26009-1-AP-20UL
This item is not returnable.
View return policy
Description
Methyltransferase that catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in various tRNAs in mitochondrion, including tRNA(Leu) (decephering codons UUA or UUG), tRNA(Lys) and tRNA(Ser) (decephering codons UCA, UCU, UCG or UCC).Specifications
TRMT61B | |
Polyclonal | |
Unconjugated | |
TRMT61B | |
TRMT61B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55006 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BVS5 | |
TRMT61B | |
TRMT61B Fusion Protein Ag23307 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRMT61B Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction