missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSP50 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12574-1-AP-150UL
This item is not returnable.
View return policy
Description
May be involved in proteolysis through its threonine endopeptidase activity.Specifications
TSP50 | |
Polyclonal | |
Unconjugated | |
PRSS50 | |
Cancer/testis antigen 20, CT20, PRSS50, Serine protease 50, testes protease 50, TSP50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235631, 29122 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BLH5, Q9UI38 | |
PRSS50 | |
TSP50 Fusion Protein Ag3247 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TSP50 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction