missing translation for 'onlineSavingsMsg'
Learn More

TTC26 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1236.20 SEK - 3313.85 SEK

Specifications

Antigen TTC26
Concentration 0.2 mg/mL
Applications Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16813625
View Documents Promotion Details
Proteintech
25083-1-AP-150UL
150 μL
3313.85 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 31-01-2022
Please to purchase this item. Need a web account? Register with us today!
16823625
View Documents Promotion Details
Proteintech
25083-1-AP-20UL
20 μL
1236.20 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 31-01-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

TTC26
Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Polyclonal
-20°C
0.2 mg/mL
Unconjugated
Rabbit
Human, Mouse
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications