missing translation for 'onlineSavingsMsg'
Learn More

Tungsten Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

433.00 SEK - 1885.00 SEK

Chemical Identifiers

CAS 7440-33-7, 7732-18-5
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
16341266
View Documents
SPEX Certiprep
PLW9-2M
30 mL Triple-leached LDPE bottle
433.00 SEK
30mL
Please to purchase this item. Need a web account? Register with us today!
10351594
View Documents
SPEX Certiprep
PLW9-2Y
125 mL Triple-leached LDPE bottle
820.00 SEK
125mL
Please to purchase this item. Need a web account? Register with us today!
10680793
View Documents
SPEX Certiprep
PLW9-2X
500 mL Triple-leached LDPE bottle
1885.00 SEK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
W1000ÎĽg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.