missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYW1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17665-1-AP-150UL
This item is not returnable.
View return policy
Specifications
TYW1B | |
Polyclonal | |
Unconjugated | |
TYW1B | |
NCRNA00069, RSAFD2, TYW1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
441250 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NUM6 | |
TYW1B | |
TYW1B Fusion Protein Ag11947 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TYW1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction