missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin (Autophagy Marker) Mouse, Clone: UBB/1748, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7314-MSM1-P1
This item is not returnable.
View return policy
Specifications
Ubiquitin (Autophagy Marker) | |
Cell Surface, Cytoplasmic and Nuclear | |
UBB/1748 | |
Unconjugated | |
Mouse | |
9kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
7314 | |
IgG2c κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human Ubiquitin protein (apprx. aa 1-119) | |
HeLa or Jurkat cells. Alzheimer s Brain | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ubiquitin (Autophagy Marker) Mouse, Clone: UBB/1748, enQuireBio™ > 100 μg
Spot an opportunity for improvement?Share a Content Correction