missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23301-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
UPF3B is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The protein is one of two functional homologs to yeast Upf3p. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome X. Two splice variants encoding different isoforms have been found for this gene.Especificaciones
UPF3B | |
Polyclonal | |
Unconjugated | |
UPF3B | |
HUPF3B, hUpf3p X, MRXS14, RENT3B, UPF3B, UPF3X | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313449, 65109, 68134 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BZI7 | |
Upf3b | |
UPF3B Fusion Protein Ag18568 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
UPF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido