missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | UPF3B |
---|---|
Concentration | 0.61 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Produktcode | Marke | Quantity | Preis | Menge & VerfĂĽgbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & VerfĂĽgbarkeit | |||||
16882275
|
Proteintech
23301-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872275
|
Proteintech
23301-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
UPF3B is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The protein is one of two functional homologs to yeast Upf3p. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome X. Two splice variants encoding different isoforms have been found for this gene.Spezifikation
UPF3B | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q9BZI7 | |
313449, 65109, 68134 | |
UPF3B Fusion Protein Ag18568 | |
Primary | |
-20°C | |
UPF3B |
0.61 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HUPF3B, hUpf3p X, MRXS14, RENT3B, UPF3B, UPF3X | |
Upf3b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
UPF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech