Learn More
Miele Professional™ Lower Basket A 151
Lower basket A 151 for holding standard DIN mesh trays as well as various inserts
Brand:  Miele Professional™ 12666340
5607.96 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15345245
  • 6175.00 SEK / Each
Estimated Shipment: 14-07-2025
Description
  • Vertical clearance dependent on upper basket selected
  • Loading width: 490 mm, depth: 435 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Miele Professional™ Lower Basket A 151 > Dimensions (D x W x H): 522 x 529 x 28mm