Learn More
Urethane, 97% Product Code.: 10430351
Urethane, 97%
Product Code. p-210718 Shop All Thermo Scientific Chemicals Products
50 g, Plastic bottle, 50g
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
Product Code. 10430351
Brand: Thermo Scientific Acros 325540500
Request Bulk or Custom Format
CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51-79-6
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00007966
InChI Key JOYRKODLDBILNP-UHFFFAOYSA-N
Synonym urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
PubChem CID 5641
ChEBI CHEBI:17967
IUPAC Name ethyl carbamate
SMILES CCOC(=O)N
Specifications
Melting Point 47.0°C to 51.0°C
Color White
Boiling Point 182.0°C to 184.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula H2NCO2C2H5
Quantity 50 g
Beilstein 03,22
Fieser 01,127
Merck Index 15,10059
Solubility Information Solubility in water: 200g/100 mL (20°C). Other solubilities: soluble in ethanol (125g/100 mL),chloroform,(111g/100 mL),ether (66.67g/100 mL),glycerol,(40g/100 mL),,slightly soluble in olive oil (3g/100 mL)
Formula Weight 89.09
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Urethane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Urethane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Product Title
Urethane, 97% >