missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USMG5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17716-1-AP-20UL
This item is not returnable.
View return policy
Description
Plays a critical role in maintaining the ATP synthase population in mitochondria.Specifications
USMG5 | |
Polyclonal | |
Unconjugated | |
USMG5 | |
HCV F transactivated protein 2, HCVFTP2, USMG5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171069, 66477, 84833 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78IK2, Q96IX5, Q9JJW3 | |
Usmg5 | |
USMG5 Fusion Protein Ag12086 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
USMG5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction