missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP1 Mouse anti-Human, Clone: 1C10H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66069-1-IG-150UL
This item is not returnable.
View return policy
Description
Deubiquitinating enzymes (DUBs) are capable of producing a free ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins in cells. DUBs possess conserved catalytic regions known as the ′cys domain′ and the ′his domain.′ There are 2 distinct families of DUBs: ubiquitin-specific processing proteases (UBPs, or USPs) and ubiquitin C-terminal hydrolases (UCHs).Specifications
USP1 | |
Monoclonal | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94782 | |
USP1 | |
USP1 Fusion Protein Ag5904 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
1C10H6 | |
Unconjugated | |
USP1 | |
Deubiquitinating enzyme 1, hUBP, ubiquitin specific peptidase 1, Ubiquitin thiolesterase 1, UBP, USP1 | |
Mouse | |
Protein A | |
RUO | |
7398 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
USP1 Mouse anti-Human, Clone: 1C10H6, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction