Learn More
BRAND™ Volumetric Flask
With glass stopper
705.00 SEK - 750.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15941322
|
BRAND™
937262 |
250 mL |
705.00 SEK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
15951322
|
BRAND™
937263 |
500 mL |
750.00 SEK
Each |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
Description
Specifications
Glass | |
Borosilicate Glass (3.3) | |
A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Volumetric Flask