Learn More
Karl Hecht™ Volumetric Flask, Conical
Designed with an enlarged base for greater stability. Karl Hecht™ Volumetric Flask, Conical is conformity certified and comes with an interchangeable stopper.
Brand:  Karl Hecht™ 42110042
 View more versions of this product
Product Code. 10309211
  • 267.00 SEK / Each
Only null left
Estimated Shipment: 20-08-2025
Description
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
Polyethylene
Conical
NS Stopper
50 mL
0.06 mL
NS 12/21
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Karl Hecht™ Volumetric Flask, Conical > Capacity: 50mL, Closure Size: NS 12/21, Tolerance: 0.06mL