Learn More
Karl Hecht™ Volumetric Flask, Conical
Designed with an enlarged base for greater stability. Karl Hecht™ Volumetric Flask, Conical is conformity certified and comes with an interchangeable stopper.
185.00 SEK - 267.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
12345793
|
Karl Hecht™
42110036 |
25 mL |
241.00 SEK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
10717132
|
Karl Hecht™
42110024 |
10 mL |
239.00 SEK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
10651162
|
Karl Hecht™
42110030 |
20 mL |
254.00 SEK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
10309211
|
Karl Hecht™
42110042 |
50 mL |
267.00 SEK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
10716752
|
Karl Hecht™
42110006 |
1 mL |
185.00 SEK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
Description
- Standard ground and interchangeable NS stopper of polyethylene.
- Conformity certified.
- Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Polyethylene | |
Conical | |
NS Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Karl Hecht™ Volumetric Flask, Conical