Learn More
Karl Hecht™ Volumetric Flask, Conical
Designed with an enlarged base for greater stability. Karl Hecht™ Volumetric Flask, Conical is conformity certified and comes with an interchangeable stopper.
Brand:  Karl Hecht™ 42110036
 View more versions of this product
Product Code. 12345793
  • 241.00 SEK / Each
Only null left
Estimated Shipment: 15-08-2025
Description
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
Polyethylene
Conical
NS Stopper
25 mL
0.04 mL
NS 10/19
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Karl Hecht™ Volumetric Flask, Conical > Capacity: 25mL, Closure Size: NS 10/19, Tolerance 0.04mL