Learn More
VPS18 Mouse anti-Human, Unconjugated, Clone: 2E10G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67590-1-IG-20UL
Description
The VPS18 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VPS18. This antibody recognizes human, rat antigen. The VPS18 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
VPS18 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2E10G11 | |
Unconjugated | |
20 μL | |
57617 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.