missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTN Mouse anti-Human, Porcine, Rabbit, Clone: 1D2C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66398-1-IG-150UL
This item is not returnable.
View return policy
Description
Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.Specifications
VTN | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04004, P22458, P48819 | |
VTN | |
VTN Fusion Protein Ag8443 | |
150 μL | |
Primary | |
Human, Pig, Rabbit | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1D2C8 | |
Unconjugated | |
VTN | |
S protein, Serum spreading factor, V75, vitronectin, VN, VNT, VTN | |
Mouse | |
Protein G | |
RUO | |
100009128, 397192, 7448 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VTN Mouse anti-Human, Porcine, Rabbit, Clone: 1D2C8, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction