Learn More
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67620-1-IG-20UL
 View more versions of this product
Product Code. 16880346
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The WASF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WASF3. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The WASF3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
WASF3
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry, Immunofluorescence
3B7D10
Unconjugated
20 μL
10810
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech > 20 μL