Learn More
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | WASF3 |
---|---|
Clone | 3B7D10 |
Concentration | 500 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880346
|
Proteintech
67620-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870346
|
Proteintech
67620-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The WASF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WASF3. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The WASF3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
WASF3 | |
500 ÎĽg/mL | |
Monoclonal | |
Mouse | |
10810 | |
Primary |
3B7D10 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse, Rat, Pig, Rabbit | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.