Learn More
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67620-1-IG-150UL
 View more versions of this product
Product Code. 16870346
  • 4652.90 SEK / 150µL
Description
The WASF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WASF3. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The WASF3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
WASF3
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry, Immunofluorescence
3B7D10
Unconjugated
150 μL
10810
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech > 150 μL