Learn More
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67620-1-IG-150UL
Description
The WASF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WASF3. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The WASF3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
WASF3 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig, Rabbit |
Western Blot, Immunohistochemistry, Immunofluorescence | |
3B7D10 | |
Unconjugated | |
150 μL | |
10810 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.