missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBSCR16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13796-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein containing regulator of chromosome condensation 1-like repeats. The encoded protein may function as a guanine nucleotide exchange factor. This gene is located in a region of chromosome 7 that is deleted in Williams-Beuren syndrome, a multisystem developmental disorder. Alternative splicing results in multiple transcript variants. Orthologs mouse allSpecifications
WBSCR16 | |
Polyclonal | |
Unconjugated | |
RCC1L | |
wbs16, WBSCR16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360796, 81554, 94254 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96I51, Q9CYF5 | |
Rcc1l | |
WBSCR16 Fusion Protein Ag4619 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WBSCR16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction