Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
3170.00 SEK - 4450.00 SEK
Specifications
For Use With Welch Compact Aspiration/Fitlration Vacuum Station
View More Specs
Products 3
Product Code Brand Pump Type Price Quantity & Availability  
Product Code Brand Pump Type Price Quantity & Availability  
10700854
View Documents
Welch™
112580-01
Hand Held Pipette
4110.00 SEK
Each
Only null left
Estimated Shipment: 07-08-2025
Add to basket
10523543
View Documents Promotion Details
Welch™
U1475K-20
Spare Receiver Kit
3170.00 SEK
Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
12759606
View Documents Promotion Details
Welch™
U1475K-21
Replacement Hydrophobic Filter
4450.00 SEK
Pack of 10
Only null left
Estimated Shipment: 01-12-2025
Add to basket
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station
Documents
Product Certifications
Product Content Correction
Product Title
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter