Learn More
WNT10B Mouse anti-Human, Unconjugated, Clone: 1G11E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67210-1-IG-20UL
 View more versions of this product
Product Code. 16889725
  • 1645.54 SEK / 20µL
Description
The WNT10B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WNT10B. This antibody recognizes human antigen. The WNT10B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
WNT10B
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1G11E4
Unconjugated
20 μL
7480
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WNT10B Mouse anti-Human, Unconjugated, Clone: 1G11E4, Proteintech > 20 μL