Learn More
WNT10B Mouse anti-Human, Unconjugated, Clone: 1G11E4, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | WNT10B |
---|---|
Clone | 1G11E4 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889725
|
Proteintech
67210-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879725
|
Proteintech
67210-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The WNT10B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WNT10B. This antibody recognizes human antigen. The WNT10B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
WNT10B | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
7480 | |
Primary |
1G11E4 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.