Learn More
WNT10B Mouse anti-Human, Unconjugated, Clone: 1G11E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67210-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The WNT10B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WNT10B. This antibody recognizes human antigen. The WNT10B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
WNT10B | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
1G11E4 | |
Unconjugated | |
150 μL | |
7480 | |
IgG1 |