missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XIAP Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23453-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.Specifications
XIAP | |
Polyclonal | |
Unconjugated | |
XIAP | |
API3, BIRC4, hIAP 3, hIAP3, hILP, IAP 3, IAP like protein, IAP3, ILP, ILP1, MIHA, X linked IAP, XIAP, XLP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
331, 63879 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P98170, Q9R0I6 | |
XIAP | |
XIAP Fusion Protein Ag0725 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
XIAP Rabbit anti-Human, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction