missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XIAP Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | XIAP |
---|---|
Concentration | 0.21 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882355
|
Proteintech
23453-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872355
|
Proteintech
23453-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.Specifications
XIAP | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat | |
P98170, Q9R0I6 | |
331, 63879 | |
XIAP Fusion Protein Ag0725 | |
Primary | |
-20°C | |
XIAP |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
API3, BIRC4, hIAP 3, hIAP3, hILP, IAP 3, IAP like protein, IAP3, ILP, ILP1, MIHA, X linked IAP, XIAP, XLP2 | |
XIAP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
XIAP Rabbit anti-Human, Rat, Polyclonal, Proteintech