missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ZDHHC12 |
---|---|
Concentration | 0.49 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884895
|
Proteintech
26433-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874895
|
Proteintech
26433-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.Specifications
ZDHHC12 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8VC90, Q96GR4 | |
66220, 84885 | |
ZDHHC12 Fusion Protein Ag23634 | |
Primary | |
-20°C | |
ZDHHC12 |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DHHC 12, ZDHHC12, Zinc finger protein 400, ZNF400 | |
ZDHHC12 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZDHHC12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech