Learn More
ZDHHC12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26433-1-AP-20UL
This item is not returnable.
View return policy
Description
ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.Specifications
ZDHHC12 | |
Polyclonal | |
Unconjugated | |
ZDHHC12 | |
DHHC 12, ZDHHC12, Zinc finger protein 400, ZNF400 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66220, 84885 | |
-20°C | |
Liquid |
Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VC90, Q96GR4 | |
ZDHHC12 | |
ZDHHC12 Fusion Protein Ag23634 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
ZDHHC12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction