missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF238 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12714-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF238 is the sequence-specific DNA-binding protein with transcriptional repression activity. ZNF238 binds the consensus DNA sequence 5′-[AC]ACATCTG[GT][AC]-3′ which contains the E box core. ZNF238 may play a role in the organization of chromosomes in the nucleus.C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.Specifications
ZNF238 | |
Polyclonal | |
Unconjugated | |
ZBTB18 | |
58 kDa repressor protein, C2H2 171, RP58, TAZ 1, TAZ1, Transcriptional repressor RP58, ZBTB18, ZFP238, zinc finger protein 238, Zinc finger protein C2H2 171, ZNF238 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10472, 30928, 64619 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99592, Q9JKY3, Q9WUK6 | |
Zbtb18 | |
ZNF238 Fusion Protein Ag3406 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF238 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction