accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11006-33-0

CAS RN 11006-33-0

IUPAC Name: 2-(2-{2-[2-(4-{2-[(6-amino-2-{1-[(2-amino-2-carbamoylethyl)amino]-2-carbamoylethyl}-5-methylpyrimidin-4-yl)formamido]-3-{[(2R,3S,4S,5S,6S)-3-{[(2S,3R,4R,5S)-4-carbamoyl-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)-3-methyloxan-2-yl]oxy}-3-(1H-imidazol-5-yl)propanamido}-3-hydroxy-2-methylpentanamido)-3-oxobutanamido]ethyl}-4,5-dihydro-1,3-thiazol-4-yl)-1,3-thiazole-4-carboxamide
Synonyms: 2-(2-{2-[2-(4-{2-[(6-amino-2-{1-[(2-amino-2-carbamoylethyl)amino]-2-carbamoylethyl}-5-methylpyrimidin-4-yl)formamido]-3-{[(2R,3S,4S,5S,6S)-3-{[(2S,3R,4R,5S)-4-carbamoyl-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)-3-methyloxan-2-yl]oxy}-3-(3H-imidazol-4-yl)propanamido}-3-hydroxy-2-methylpentanamido)-3-oxobutanamido]ethyl}-4,5-dihydro-1,3-thiazol-4-yl)-1,3-thiazole-4-carboxamide
Molecular Weight (g/mol): 1326.38
Molecular Formula: C51H75N17O21S2
InChi Key: QRBLKGHRWFGINE-UGWAGOLRSA-N
SMILES: CC(NC(=O)C(NC(=O)C1=NC(=NC(N)=C1C)C(CC(N)=O)NCC(N)C(N)=O)C(O[C@@H]1O[C@@H](CO)[C@@H](O)[C@H](O)[C@]1(C)O[C@@H]1OC(CO)[C@H](O)[C@@](O)([C@H]1O)C(N)=O)C1=CN=CN1)C(O)C(C)C(=O)NC(C(C)=O)C(=O)NCCC1=NC(CS1)C1=NC(=CS1)C(N)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Type 
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products