accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 110871-86-8

H3C(S)N(R)CH3HNFNH2ONFOHO

CAS RN 110871-86-8

IUPAC Name: 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: sparfloxacin zagam sparfloxacinum sparfloxacine esparfloxacino SPFX
Molecular Weight (g/mol): 392.41
Molecular Formula: C19H22F2N4O3
InChi Key: DZZWHBIBMUVIIW-DTORHVGOSA-N
SMILES: C[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products