accessibility menu, dialog, popup

UserName

CAS RN 111-44-4

ClOCl

CAS RN 111-44-4

IUPAC Name: 1-chloro-2-(2-chloroethoxy)ethane
Synonyms: BCEE 2,2'-dicloroetiletere caswell no. 309 2,2'-dichlorodiethyl ether epa pesticide chemical code 029501 2,2'-dichlor-diaethylaether bis(chloro-2-ethyl) oxide clorex β,β-dichlorodiethyl ether ccris 88
Molecular Weight (g/mol): 143.01
Molecular Formula: C4H8Cl2O
InChi Key: ZNSMNVMLTJELDZ-UHFFFAOYSA-N
SMILES: ClCCOCCCl
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results