CAS RN 1111556-37-6
CAS RN 1111556-37-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris™ Takinib

Potent and selective TAK1/MAP3K7 kinase inhibitor
2
Medchem Express Takinib, MedChemExpress
MedChemExpress Takinib (EDHS-206) is an orally active and selective TAK1 inhibitor (IC50=9.5 nM), more than 1.5 log more potent than the second and third ranked targets, IRAK4 (120 nM) and IRAK1 (390 nM), respectively.
